Cyclopropyl(3,5-dichloro-2-fluorophenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

478008-5G 478008-25G 478008-1G 478008-500MG
77629-762EA 1484.23 USD
77629-762 77635-942 77643-204 77647-198
Cyclopropyl(3,5-dichloro-2-fluorophenyl)methanone ≥95%
Cyclopropyl(3,5-dichloro-2-fluorophenyl)methanone

Precaución: For Research Use Only

Formula: C₁₀H₇Cl₂FO
Storage Temperature: Ambient
CAS Number: 3031804-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR