1-(3,5-Dichloro-2-fluorophenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

478007-5G 478007-25G 478007-1G 478007-500MG
77629-760EA 1484.23 USD
77629-760 77659-650 77643-202 77651-162
1-(3,5-Dichloro-2-fluorophenyl)propan-1-one ≥95%
1-(3,5-Dichloro-2-fluorophenyl)propan-1-one

Precaución: For Research Use Only

Formula: C₉H₇Cl₂FO
Storage Temperature: Ambient
CAS Number: 3031789-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR