(3,5-Dichloro-2-fluorophenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

478006-5G 478006-1G 478006-25G 478006-500MG
77633-334EA 1441 USD
77633-334 77639-162 77659-648 77651-160
(3,5-Dichloro-2-fluorophenyl)trimethylsilane ≥95%
(3,5-Dichloro-2-fluorophenyl)trimethylsilane

Precaución: For Research Use Only

Formula: 237.174 g/mol
MW: C₉H₁₁Cl₂FSi
Storage Temperature: Ambient
CAS Number: 3031809-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR