Tert-butyl 5-chloro-2-fluoro-3-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

477963-5G 477963-25G 477963-1G 477963-500MG
77629-756EA 1484.23 USD
77629-756 77635-940 77643-198 77647-194
Tert-butyl 5-chloro-2-fluoro-3-methylbenzoate ≥95%
Tert-butyl 5-chloro-2-fluoro-3-methylbenzoate

Precaución: For Research Use Only

Formula: 244.69 g/mol
MW: C₁₂H₁₄ClFO₂
Storage Temperature: Ambient
CAS Number: 3031784-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR