(5-Chloro-2-fluoro-3-methylphenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

477962-5G 477962-25G 477962-1G 477962-500MG
77629-754EA 1484.23 USD
77629-754 77656-076 77643-196 77651-156
(5-Chloro-2-fluoro-3-methylphenyl)trimethylsilane ≥97%
(5-Chloro-2-fluoro-3-methylphenyl)trimethylsilane

Precaución: For Research Use Only

Formula: C₁₀H₁₄ClFSi
Storage Temperature: Ambient
CAS Number: 3031789-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR