1-(5-Chloro-2-fluoro-3-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

477960-5G 477960-25G 477960-1G 477960-500MG
77629-752EA 1484.23 USD
77629-752 77659-642 77643-194 77651-154
1-(5-Chloro-2-fluoro-3-methylphenyl)cyclobutanol ≥95%
1-(5-Chloro-2-fluoro-3-methylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₁H₁₂ClFO
Storage Temperature: Ambient
CAS Number: 3031813-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR