1-(5-Chloro-2-fluoro-3-methylphenyl)-2-methoxyethanone ≥95%

Supplier: AOBChem USA

Warning

477959-5G 477959-1G 477959-25G 477959-500MG
77629-750EA 1484.23 USD
77629-750 77639-158 77659-640 77647-192
1-(5-Chloro-2-fluoro-3-methylphenyl)-2-methoxyethanone ≥95%
1-(5-Chloro-2-fluoro-3-methylphenyl)-2-methoxyethanone

Precaución: For Research Use Only

Formula: 216.637 g/mol
MW: C₁₀H₁₀ClFO₂
Storage Temperature: Ambient
CAS Number: 3031789-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR