5-Bromo-6-methoxy-2-naphthaldehyde ≥97%

Supplier: AOBChem USA

Warning

47751-5G 47751-1G 47751-25G 47751-500MG
77633-330EA 1115 USD
77633-330 77639-154 77659-636 77647-188
5-Bromo-6-methoxy-2-naphthaldehyde ≥97%
5-Bromo-6-methoxy-2-naphthaldehyde

Precaución: For Research Use Only

Formula: C₁₂H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD22192769
CAS Number: 148248-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR