5-Bromo-6-methoxyquinoline ≥97%

Supplier: AOBChem USA

Warning

47750-5G 47750-1G 47750-25G 47750-500MG
77633-328EA 827 USD
77633-328 77639-152 77659-634 77647-186
5-Bromo-6-methoxyquinoline ≥97%
5-Bromo-6-methoxyquinoline

Precaución: For Research Use Only

Formula: C₁₀H₈BrNO
MW: 238.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18452262
CAS Number: 36023-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR