2-(4-Chlorophenyl)-3-nitropyridine ≥97%

Supplier: AOBChem USA

Warning

477266-5G 477266-1G 477266-500MG 477266-25G
77633-326EA 894 USD
77633-326 77639-150 77652-886 77659-632
2-(4-Chlorophenyl)-3-nitropyridine ≥97%
2-(4-Chlorophenyl)-3-nitropyridine

Precaución: For Research Use Only

Formula: C₁₁H₇ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06200957
CAS Number: 886361-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR