1-(2-Bromo-5-(trifluoromethyl)phenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

47720-1G 47720-5G 47720-500MG
77635-072EA 830 USD
77635-072 77659-628 77647-182
1-(2-Bromo-5-(trifluoromethyl)phenyl)ethanol ≥95%
1-(2-Bromo-5-(trifluoromethyl)phenyl)ethanol

Precaución: For Research Use Only

Formula: C₉H₈BrF₃O
MW: 269.058 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26137983
CAS Number: 1443376-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR