3,4-Diisopropoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

477063-5G 477063-1G 477063-25G 477063-500MG
77629-746EA 1484.23 USD
77629-746 77639-148 77659-626 77647-180
3,4-Diisopropoxybenzamide ≥97%
3,4-Diisopropoxybenzamide

Precaución: For Research Use Only

Formula: C₁₃H₁₉NO₃
Storage Temperature: Ambient
CAS Number: 1369817-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR