3,4-Diisopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

477061-1G 477061-5G 477061-500MG
77639-146EA 657 USD
77639-146 77656-074 77651-152
3,4-Diisopropoxybenzaldehyde ≥97%
3,4-Diisopropoxybenzaldehyde

Precaución: For Research Use Only

Formula: C₁₃H₁₈O₃
Storage Temperature: Ambient
CAS Number: 64000-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR