1-(4-(Cyclopentyloxy)phenyl)-2-methoxyethanone ≥95%

Supplier: AOBChem USA

Warning

476719-5G 476719-1G 476719-25G 476719-500MG
77629-740EA 1484.23 USD
77629-740 77639-142 77656-072 77647-174
1-(4-(Cyclopentyloxy)phenyl)-2-methoxyethanone ≥95%
1-(4-(Cyclopentyloxy)phenyl)-2-methoxyethanone

Precaución: For Research Use Only

Formula: 234.2909 g/mol
MW: C₁₄H₁₈O₃
Storage Temperature: Ambient
CAS Number: 3031799-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR