4-Bromo-2'-methylbenzophenone ≥95%

Supplier: AOBChem USA

Warning

47659-5G 47659-25G 47659-1G 47659-500MG
77635-070EA 342 USD
77635-070 77656-070 77647-170 77647-172
4-Bromo-2'-methylbenzophenone ≥95%
4-Bromo-2'-methylbenzophenone

Precaución: For Research Use Only

Formula: C₁₄H₁₁BrO
MW: 275.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801661
CAS Number: 27428-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR