2-Bromodiphenylmethane ≥95%

Supplier: AOBChem USA

Warning

47648-5G 47648-25G
77635-068EA 107 USD
77635-068 77656-068
2-Bromodiphenylmethane ≥95%
2-Bromodiphenylmethane

Precaución: For Research Use Only

Formula: C₁₃H₁₁Br
MW: 247.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00079713
CAS Number: 23450-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR