7-Bromo-3-cyclopropyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

476128-1G 476128-5G 476128-500MG
77639-140EA 574 USD
77639-140 77656-064 77651-146
7-Bromo-3-cyclopropyl-1H-indazole ≥97%
7-Bromo-3-cyclopropyl-1H-indazole

Precaución: For Research Use Only

Formula: C₁₀H₉BrN₂
Storage Temperature: Ambient
CAS Number: 1360927-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR