1-(3,5-Di-tert-butylphenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

476006-5G 476006-25G 476006-1G 476006-500MG
77629-736EA 1484.23 USD
77629-736 77659-620 77643-182 77647-166
1-(3,5-Di-tert-butylphenyl)propan-1-ol ≥95%
1-(3,5-Di-tert-butylphenyl)propan-1-ol

Precaución: For Research Use Only

Formula: 248.404 g/mol
MW: C₁₇H₂₈O
Storage Temperature: Ambient
CAS Number: 3031799-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR