1-(3,5-Di-tert-butylphenyl)-2-methoxyethanone ≥95%

Supplier: AOBChem USA

Warning

476005-1G 476005-5G 476005-500MG
77639-138EA 560 USD
77639-138 77656-062 77647-164
1-(3,5-Di-tert-butylphenyl)-2-methoxyethanone ≥95%
1-(3,5-Di-tert-butylphenyl)-2-methoxyethanone

Precaución: For Research Use Only

Formula: C₁₇H₂₆O₂
Storage Temperature: Ambient
CAS Number: 1256481-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR