Tert-butyl 3,5-di-tert-butylbenzoate ≥95%

Supplier: AOBChem USA

Warning

476003-5G 476003-25G 476003-1G 476003-500MG
77629-734EA 1484.23 USD
77629-734 77635-930 77643-180 77651-144
Tert-butyl 3,5-di-tert-butylbenzoate ≥95%
Tert-butyl 3,5-di-tert-butylbenzoate

Precaución: For Research Use Only

Formula: C₁₉H₃₀O₂
Storage Temperature: Ambient
CAS Number: 3009009-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR