1-(2,6-Difluoro-3-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

476000-5G 476000-25G 476000-1G 476000-500MG
77629-732EA 1484.23 USD
77629-732 77656-060 77643-178 77651-142
1-(2,6-Difluoro-3-methylphenyl)cyclobutanol ≥95%
1-(2,6-Difluoro-3-methylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₁H₁₂F₂O
Storage Temperature: Ambient
CAS Number: 1548129-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR