1-(2-Fluoro-5-iodophenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

475984-5G 475984-1G 475984-500MG
77629-730EA 1818.98 USD
77629-730 77643-176 77647-162
1-(2-Fluoro-5-iodophenyl)ethanol ≥95%
1-(2-Fluoro-5-iodophenyl)ethanol

Precaución: For Research Use Only

Formula: C₈H₈FIO
Storage Temperature: Ambient
CAS Number: 2386021-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR