2-(2-Fluoro-5-iodophenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

475983-5G 475983-1G 475983-25G 475983-500MG
77633-318EA 1312 USD
77633-318 77639-134 77656-058 77647-160
2-(2-Fluoro-5-iodophenyl)propan-2-ol ≥95%
2-(2-Fluoro-5-iodophenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 280.078 g/mol
MW: C₉H₁₀FIO
Storage Temperature: Ambient
CAS Number: 1935904-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR