1-(2-Fluoro-5-iodophenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

475982-5G 475982-25G 475982-1G 475982-500MG
77629-728EA 1484.23 USD
77629-728 77656-056 77643-174 77647-158
1-(2-Fluoro-5-iodophenyl)propan-1-ol ≥95%
1-(2-Fluoro-5-iodophenyl)propan-1-ol

Precaución: For Research Use Only

Formula: 280.078 g/mol
MW: C₉H₁₀FIO
Storage Temperature: Ambient
CAS Number: 3031805-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR