3-(2-Fluoro-5-iodophenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

475978-5G 475978-1G 475978-25G 475978-500MG
77629-726EA 1484.23 USD
77629-726 77639-130 77656-052 77651-138
3-(2-Fluoro-5-iodophenyl)oxetan-3-ol ≥95%
3-(2-Fluoro-5-iodophenyl)oxetan-3-ol

Precaución: For Research Use Only

Formula: C₉H₈FIO₂
Storage Temperature: Ambient
CAS Number: 3031788-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR