Methyl 2-(2-fluoro-5-iodophenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

475977-5G 475977-25G 475977-1G 475977-500MG
77629-724EA 1484.23 USD
77629-724 77635-928 77643-172 77651-136
Methyl 2-(2-fluoro-5-iodophenyl)-2-oxoacetate ≥95%
Methyl 2-(2-fluoro-5-iodophenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 308.045 g/mol
MW: C₉H₆FIO₃
Storage Temperature: Ambient
CAS Number: 3031789-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR