(3-Chloro-2-fluorophenyl)(cyclohexyl)methanone ≥95%

Supplier: AOBChem USA

Warning

475975-5G 475975-25G 475975-1G 475975-500MG
77633-316EA 1381 USD
77633-316 77659-616 77647-154 77651-134
(3-Chloro-2-fluorophenyl)(cyclohexyl)methanone ≥95%
(3-Chloro-2-fluorophenyl)(cyclohexyl)methanone

Precaución: For Research Use Only

Formula: 240.701 g/mol
MW: C₁₃H₁₄ClFO
Storage Temperature: Ambient
CAS Number: 1342629-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR