Cyclobutyl(2,6-difluoro-3-iodophenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

475964-5G 475964-25G 475964-1G 475964-500MG
77629-720EA 1484.23 USD
77629-720 77635-926 77643-168 77647-150
Cyclobutyl(2,6-difluoro-3-iodophenyl)methanone ≥95%
Cyclobutyl(2,6-difluoro-3-iodophenyl)methanone

Precaución: For Research Use Only

Formula: C₁₁H₉F₂IO
Storage Temperature: Ambient
CAS Number: 3031789-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR