Cyclopropyl(2,6-difluoro-3-iodophenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

475963-5G 475963-25G 475963-1G 475963-500MG
77629-718EA 1484.23 USD
77629-718 77635-924 77643-166 77651-130
Cyclopropyl(2,6-difluoro-3-iodophenyl)methanone ≥95%
Cyclopropyl(2,6-difluoro-3-iodophenyl)methanone

Precaución: For Research Use Only

Formula: C₁₀H₇F₂IO
Storage Temperature: Ambient
CAS Number: 3031805-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR