1-(2,6-Difluoro-3-iodophenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

475962-5G 475962-25G 475962-1G 475962-500MG
77629-716EA 1484.23 USD
77629-716 77656-048 77643-164 77651-128
1-(2,6-Difluoro-3-iodophenyl)propan-1-one ≥95%
1-(2,6-Difluoro-3-iodophenyl)propan-1-one

Precaución: For Research Use Only

Formula: 296.053 g/mol
MW: C₉H₇F₂IO
Storage Temperature: Ambient
CAS Number: 3031804-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR