1-(2,6-Difluoro-3-iodophenyl)-2-methoxyethanone ≥95%

Supplier: AOBChem USA

Warning

475961-5G 475961-1G 475961-25G 475961-500MG
77629-714EA 1484.23 USD
77629-714 77639-126 77659-612 77651-126
1-(2,6-Difluoro-3-iodophenyl)-2-methoxyethanone ≥95%
1-(2,6-Difluoro-3-iodophenyl)-2-methoxyethanone

Precaución: For Research Use Only

Formula: 312.052 g/mol
MW: C₉H₇F₂IO₂
Storage Temperature: Ambient
CAS Number: 3031789-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR