4-(2,6-Difluorophenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

475957-5G 475957-1G 475957-500MG
77629-712EA 1484.23 USD
77629-712 77639-124 77651-124
4-(2,6-Difluorophenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(2,6-Difluorophenyl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: 214.209 g/mol
MW: C₁₁H₁₂F₂O₂
Storage Temperature: Ambient
CAS Number: 1341452-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR