1-(2,6-Difluorophenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

475955-5G 475955-25G 475955-1G 475955-500MG
77633-312EA 1312 USD
77633-312 77659-610 77643-162 77647-146
1-(2,6-Difluorophenyl)propan-1-ol ≥95%
1-(2,6-Difluorophenyl)propan-1-ol

Precaución: For Research Use Only

Formula: C₉H₁₀F₂O
Storage Temperature: Ambient
MDL Number: MFCD19601748
CAS Number: 1339492-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR