1-(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

475953-5G 475953-1G 475953-25G 475953-500MG
77629-710EA 1484.23 USD
77629-710 77639-122 77656-044 77651-120
1-(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)propan-1-ol ≥95%
1-(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)propan-1-ol

Precaución: For Research Use Only

Formula: 301.075 g/mol
MW: C₁₀H₉BrF₄O
Storage Temperature: Ambient
CAS Number: 3031793-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR