3-(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

475952-5G 475952-1G 475952-25G 475952-500MG
77629-708EA 1484.23 USD
77629-708 77639-120 77656-042 77647-144
3-(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)oxetan-3-ol ≥95%
3-(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)oxetan-3-ol

Precaución: For Research Use Only

Formula: C₁₀H₇BrF₄O₂
MW: 315.059 g/mol
Storage Temperature: Ambient
CAS Number: 3031809-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR