(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(cyclobutyl)methanone ≥95%

Supplier: AOBChem USA

Warning

475951-5G 475951-25G 475951-1G 475951-500MG
77629-706EA 1519.25 USD
77629-706 77656-040 77643-160 77647-142
(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(cyclobutyl)methanone ≥95%
(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(cyclobutyl)methanone

Precaución: For Research Use Only

Formula: 325.097 g/mol
MW: C₁₂H₉BrF₄O
Storage Temperature: Ambient
CAS Number: 3031789-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR