(3-(Benzyloxy)-5-methylphenyl)(cyclopropyl)methanone ≥95%

Supplier: AOBChem USA

Warning

475948-5G 475948-500MG 475948-25G 475948-1G
77633-310EA 1441 USD
77633-310 77652-884 77659-606 77643-158
(3-(Benzyloxy)-5-methylphenyl)(cyclopropyl)methanone ≥95%
(3-(Benzyloxy)-5-methylphenyl)(cyclopropyl)methanone

Precaución: For Research Use Only

Formula: C₁₈H₁₈O₂
Storage Temperature: Ambient
CAS Number: 3031805-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR