1-(3-(Benzyloxy)-5-methylphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

475944-5G 475944-25G 475944-1G 475944-500MG
77629-700EA 1484.23 USD
77629-700 77659-604 77643-154 77647-136
1-(3-(Benzyloxy)-5-methylphenyl)propan-1-one ≥95%
1-(3-(Benzyloxy)-5-methylphenyl)propan-1-one

Precaución: For Research Use Only

Formula: 254.324 g/mol
MW: C₁₇H₁₈O₂
Storage Temperature: Ambient
CAS Number: 3031798-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR