(4-Chloro-2-fluoro-3-methylphenyl)(cyclobutyl)methanone ≥95%

Supplier: AOBChem USA

Warning

475943-5G 475943-1G 475943-500MG
77659-602EA 2073 USD
77659-602 77643-152 77647-134
(4-Chloro-2-fluoro-3-methylphenyl)(cyclobutyl)methanone ≥95%
(4-Chloro-2-fluoro-3-methylphenyl)(cyclobutyl)methanone

Precaución: For Research Use Only

Formula: C₁₂H₁₂ClFO
Storage Temperature: Ambient
CAS Number: 1870344-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR