1-(4-Chloro-2-fluoro-3-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

475942-1G 475942-5G 475942-500MG
77639-118EA 580 USD
77639-118 77656-036 77651-116
1-(4-Chloro-2-fluoro-3-methylphenyl)cyclobutanol ≥95%
1-(4-Chloro-2-fluoro-3-methylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 214.664 g/mol
MW: C₁₁H₁₂ClFO
Storage Temperature: Ambient
CAS Number: 1874608-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR