5-Bromo-2-ethyl-4-methylthiazole ≥97%
Supplier: AOBChem USA
|
Warning
|
475825-5G
475825-25G
77635-066EA
141
USD
77635-066
77656-034
5-Bromo-2-ethyl-4-methylthiazole ≥97%
5-Bromo-2-ethyl-4-methylthiazole
Precaución: For Research Use Only
Formula:
C₆H₈BrNS MW: 206.10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18311867 CAS Number: 863190-90-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...