5-Bromo-2-(1-bromoethyl)-1,3-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

475707-5G 475707-25G 475707-1G 475707-500MG
77629-692EA 1484.23 USD
77629-692 77635-920 77643-148 77651-110
5-Bromo-2-(1-bromoethyl)-1,3-difluorobenzene ≥97%
5-Bromo-2-(1-bromoethyl)-1,3-difluorobenzene

Precaución: For Research Use Only

Formula: C₈H₆Br₂F₂
Storage Temperature: Ambient
CAS Number: 1935074-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR