2-Chloro-1,4-difluoro-3-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

475683-5G 475683-1G 475683-25G 475683-500MG
77635-064EA 542 USD
77635-064 77639-112 77659-600 77647-126
2-Chloro-1,4-difluoro-3-iodobenzene ≥97%
2-Chloro-1,4-difluoro-3-iodobenzene

Precaución: For Research Use Only

Formula: C₆H₂ClF₂I
MW: 274.434 g/mol
Storage Temperature: Ambient
CAS Number: 1208077-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR