Ethyl 4-(tert-butyl)thiazole-2-carboxylate ≥95%

Supplier: AOBChem USA

Warning

475648-5G 475648-25G 475648-1G 475648-500MG
77635-062EA 399 USD
77635-062 77656-030 77643-144 77647-124
Ethyl 4-(tert-butyl)thiazole-2-carboxylate ≥95%
Ethyl 4-(tert-butyl)thiazole-2-carboxylate

Precaución: For Research Use Only

Formula: C₁₀H₁₅NO₂S
MW: 213.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14654431
CAS Number: 79247-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR