5-Bromo-2,4-dimethyl-1,3-thiazole ≥97%

Supplier: AOBChem USA

Warning

475253-5G 475253-25G
77635-060EA 88 USD
77635-060 77656-028
5-Bromo-2,4-dimethyl-1,3-thiazole ≥97%
5-Bromo-2,4-dimethyl-1,3-thiazole

Precaución: For Research Use Only

Formula: C₅H₆BrNS
MW: 192.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03086099
CAS Number: 28599-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR