Cyclopropyl(4,5-dimethyl-2-thiazolyl)methanone ≥95%

Supplier: AOBChem USA

Warning

475217-5G 475217-1G 475217-25G 475217-500MG
77633-306EA 1154 USD
77633-306 77639-110 77656-026 77647-122
Cyclopropyl(4,5-dimethyl-2-thiazolyl)methanone ≥95%
Cyclopropyl(4,5-dimethyl-2-thiazolyl)methanone

Precaución: For Research Use Only

Formula: 181.26 g/mol
MW: C₉H₁₁NOS
Storage Temperature: Ambient
CAS Number: 1211514-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR