2-Chloro-4,5-dimethyl-1,3-thiazole ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-4,5-dimethylthiazole

475188-5G 475188-1G 475188-25G 475188-500MG
77635-058EA 419 USD
77635-058 77639-106 77659-596 77647-118
2-Chloro-4,5-dimethyl-1,3-thiazole ≥95%
2-Chloro-4,5-dimethyl-1,3-thiazole

Precaución: For Research Use Only

Formula: C₅H₆ClNS
MW: 147.626 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17249933
CAS Number: 56355-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR