2-Fluoro-3-methyl-4-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

47449-1G 47449-5G 47449-500MG
77635-056EA 957 USD
77635-056 77659-594 77647-116
2-Fluoro-3-methyl-4-(trifluoromethyl)benzaldehyde ≥97%
2-Fluoro-3-methyl-4-(trifluoromethyl)benzaldehyde

Precaución: For Research Use Only

Formula: 206.137 g/mol
MW: C₉H₆F₄O
Storage Temperature: Ambient
MDL Number: MFCD34813625
CAS Number: 2384515-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR