5-Bromo-2-fluoronicotinonitrile ≥97%

Supplier: AOBChem USA

Warning

47410-5G 47410-25G 47410-1G 47410-500MG
77635-054EA 252 USD
77635-054 77656-018 77643-136 77651-100
5-Bromo-2-fluoronicotinonitrile ≥97%
5-Bromo-2-fluoronicotinonitrile

Precaución: For Research Use Only

Formula: C₆H₂BrFN₂
Storage Temperature: Ambient
MDL Number: MFCD18258637
CAS Number: 1256821-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR