2-(Benzyloxy)-3-bromo-5-fluoropyridine ≥95%

Supplier: AOBChem USA

Warning

473674-5G 473674-25G 473674-1G 473674-500MG
77633-300EA 688 USD
77633-300 77659-592 77643-134 77647-114
2-(Benzyloxy)-3-bromo-5-fluoropyridine ≥95%
2-(Benzyloxy)-3-bromo-5-fluoropyridine

Precaución: For Research Use Only

Formula: C₁₂H₉BrFNO
Storage Temperature: Ambient
CAS Number: 1206776-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR